Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
Actual product may vary
Fiberlite™ F13-14 x 50cy Fixed Angle Rotor
Execute a complete workflow in a single conical tube, achieving time and cost efficiencies as well as waste reduction with this fixed angle rotor.
Have Questions?
Catalog NumberFor Use With (Equipment)
46922Thermo Scientific Sorvall RC 6 Plus Centrifuge
Catalog number 46922
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Thermo Scientific Sorvall RC 6 Plus Centrifuge
Support a complete workflow in single conical tube with the Thermo Scientific™ Fiberlite™ F13-14 x 50cy Fixed Angle Rotor, which enables time and cost efficiencies as well as waste reduction.
Features:
  • Reduce run times with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design for structural integrity
  • Backed by a 15-year warranty
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Compatible with:
Beckman™ J2 Series, Avanti Series (except Avanti J-HC), Thermo Scientific Sorvall™ RC 6 Plus Series, Jouan KR22i and Hitachi™ Himac™ CR21/CR22 Series centrifuges.
Recommended for:
Subcellular fractions, bacteria pelleting, lysate clarification, cell membranes, sucrose gradient isolution of large sub cellular particles and gradient separation of cells.
Note:
†Performance may vary by centrifuge model.
Specifications
DescriptionFiberlite F13-14 x 50cy Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity4 x 50 mL
Max. RCF29,000 x g
Max. Speed13,000 rpm
Radius (Metric)153 mm
Radius (Metric) Min.83 mm
K Factor916
Max. Tube Length115 mm
Max. Tube Diameter29 mm
Net Weight15.5 lb. (7.03 kg)
For Use With (Equipment)Thermo Scientific Sorvall RC 6 Plus Centrifuge
Angle34°
Certifications/ComplianceBiocontainment Certified by CAMR, HPA, Porton Down, U.K.
Min. RCF15,710 x g
BiocontainmentYes
Green FeaturesEnergy efficient, less waste
Unit SizeEA