Finnpipette™ F2 Multichannel Pipettes
Actual product may vary
Thermo Scientific
Finnpipette™ F2 Multichannel Pipettes
Use this multichannel pipette for highly demanding applications where robustness and durability are essential.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of Channels
466203030 to 300 μL8
46620001 to 10 μL8
46620105 to 50 μL8
466202010 to 100 μL8
46620401 to 10 μL12
46620505 to 50 μL12
466206010 to 100 μL12
466207030 to 300 μL12
46620801 to 10 μL16
46620905 to 50 μL16
Catalog number 4662030
HSN 84798999
Price (SGD)
1,409.00
Each
Volume (Metric):
30 to 300 μL
Number of Channels:
8
Price (SGD)
1,409.00
Each
Use the Thermo Scientific™ Finnpipette™ F2 Multichannel Pipette for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, this pipette is built with PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable and available in 8-, 12- and 16-channel models with a selection of volume ranges. In the low volume models, the super blow-out function ensures accurate dispensing for even the lowest volumes.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorOrange
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Unit SizeEach