Finnpipette™ F1 Multichannel Pipettes
Actual product may vary
Thermo Scientific
Finnpipette™ F1 Multichannel Pipettes
Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of ChannelsColor
4661090N5 to 50 μL16Turquoise
4661000N1 to 10 μL8Pink
4661010N5 to 50 μL8Yellow
4661020N10 to 100 μL8Yellow
4661030N30 to 300 μL8Orange
4661040N1 to 10 μL12Pink
4661050N5 to 50 μL12Yellow
4661060N10 to 100 μL12Yellow
4661070N30 to 300 μL12Orange
4661080N1 to 10 μL16Purple
Catalog number 4661090N
HSN 84798999
Price (SGD)
1,609.00
Special offer
Ends: 30-Jun-2026
1,787.00
Save 178.00 (10%)
Each
Volume (Metric):
5 to 50 μL
Number of Channels:
16
Color:
Turquoise
Price (SGD)
1,609.00
Special offer
Ends: 30-Jun-2026
1,787.00
Save 178.00 (10%)
Each
Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette F1
ColorTurquoise
Number of Channels16
Volume (Metric)5 to 50 μL
Increments0.1μL
Unit SizeEach