Finnpipette™ Novus Multichannel Pipettes
Finnpipette™ Novus Multichannel Pipettes
Actual product may vary
Thermo Scientific
Finnpipette™ Novus Multichannel Pipettes
Speed up repetitive dispensing into microplates with these lightweight, multichannel pipettes, with balanced weight distribution for reduced muscle stress and strain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Number of ChannelsColor
46300800100 to 1200 μL8Turquoise
463002005 to 50 μL8Yellow
463003005 to 50 μL12Yellow
4630040030 to 300 μL8Orange
4630050030 to 300 μL12Orange
463007005 to 50 μL16Turquoise
FIS21-377-845100 to 1200 μL8Turquoise
FKS11722245100 to 1200 μL8Turquoise
Catalog number 46300800
Price (SGD)
2,085.00
EA
Add to cart
Volume (Metric):
100 to 1200 μL
Number of Channels:
8
Color:
Turquoise
Price (SGD)
2,085.00
EA
Add to cart
Speed up repetitive dispensing into 96- and 384-microplates with Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes. Available in a wide range of models for common laboratory tasks, this product has a lightweight handle and unique index-finger operation that enable longer work periods without fatigue. Simplicity, comfort and functionality — the Novus will be your new best friend in the lab.
Features:
  • Easy-to-read and intuitive menu navigation
  • Multilingual user interface options
  • 10 pipetting options include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more
    • 9 aspirate/dispense speeds 
    • Save up to 9 programs with your most common protocols
    • 8 multichannel versions, including a 16-channel for 384-well microplate applications, with color coding by volume for easy recognition
    • Full-size, backlit screen eliminates surrounding light reflections and improves contrast in low-light conditions
    • Quick in-lab calibration for accuracy and precision
    • Fully autoclavable tip cone ensures sterility and avoids cross-contamination
    • Long-life battery allows approximately 4000 pipetting operations; recharged in about one hour
Ergonomic Design
  • Extremely lightweight construction
  • Trigger-action pipetting — index finger controls dispensing with just a soft press of a button
  • 120° adjustable finger rest for balance and comfort
  • Soft-touch tip ejection minimizes thumb strain; can be operated by either hand
Compatible with:
Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips
Includes:
Finnpipette Novus, universal charger, piston removal tool, instructions for use and calibration certificate
Specifications
Accuracy3.0% ±1.0
Compatible TipsFlex 1200
AutoclavableAutoclavable
ColorTurquoise
For Use WithFlex 1200
ModelNovus
Number of Channels8
Product LineNovus
ProgrammabilityYes
SterilitySterile
TypeMulti-Channel Pipette
Volume (Metric)100 to 1200 μL
Increments1 μL
Precision0.9 to 0.2%
Unit SizeEA