S100-AT4 Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
S100-AT4 Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Actual product may vary
S100-AT4 Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Support capacities of 6 x 3.5mL, with additional support for microtubes at speeds up to 100,000rpm.
Have Questions?
Catalog NumberCapacity
455866 x 3.5 mL
Catalog number 45586
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
6 x 3.5 mL
Support capacities of 6 x 3.5mL, with additional support for microtubes at speeds up to 100,000rpm. The Thermo Scientific™ S100-AT4 Fixed Angle Rotor can also support microtubes.
Features:
  • Fixed angle (30°) titanium rotor
  • Capacity: 6 × 3.5mL
  • Max. Speed/RCF: 100,000rpm/540,628 x g
  • K factor: 15.9
Compatible with:
Thermo Scientific Sorvall MTX 150 Micro-Ultracentrifuge series, Sorvall MX Series Floor Model Micro-Ultracentrifuge
Recommended for:
Small volume protein virus and DNA isolation.
Specifications
DescriptionS100-AT4 Fixed Angle Rotor
MaterialTitanium
TypeFixed Angle Rotor
Capacity6 x 3.5 mL
Max. RCF540,628 x g
Max. Speed100,000 rpm
Radius (Metric)48.4 mm
Radius (Metric) Min.25.8 mm
K Factor15.9
Max. Tube Length51 mm
Max. Tube Diameter13 mm
Net Weight2.4 lb. (1.07 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
Angle30°
Avg. RCF414,407 x g
Min. RCF288,186 x g
Unit SizeEA