Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Actual product may vary
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Comfortably perform daily pipetting tasks with these reliable and durable manual variable volume pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color Code
4500100
also known as 4500100P
5 to 50 μLOrange
450-00200.5 to 10 μLGray
Catalog number 4500100
also known as 4500100P
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5 to 50 μL
Color Code:
Orange
Thermo Scientific™ Finnpipette™ Digital Variable Volume Single Channel Pipettes feature a classic style for timeless dependability and durability. The Finnpipette Digital pipette is fully autoclavable, and combines robust durability with consistent performance. Finnpipette Digital pipettes are available in single and multichannel models.
Consistent Performance
  • Fully autoclavable
  • No metal parts to rust or corrode
  • Delivers micro-size drops with the patented super blow-out piston in 50μL models and below
Comfort and Ease of Use
  • Easily set volume by t urning the large operating button
  • Quickly read volume selection in large digital window display
  • Gearing mechanism releases tips with a gentle push
Specifications
Accuracy±3 to 0.6%
Compatible TipsFinnitip Flex 200, 200 Ext, 250 Univ, QSP tip 200, 300
AutoclavableAutoclavable
ColorOrange
For Use WithFinnTip™:Flex 200, 200 Extended, 250 Universal, QSP™ Tip:200, 300
ModelFinnpipette™ Digital
Number of Channels1
Product LineFinnpipette
SterilitySterile
TypePipette
Volume (Metric)5 to 50 μL
Increments0.5 μL
Precision2.5 to 0.3%
Color CodeOrange
Unit SizeEA