MICRO-TOUCH™ NITRA-TEX™ Nitrile Examination Gloves
MICRO-TOUCH™ NITRA-TEX™ Nitrile Examination Gloves
Actual product may vary
MICRO-TOUCH™ NITRA-TEX™ Nitrile Examination Gloves
Ansell MICRO-TOUCH™ NITRA-TEX™ Nitrile Examination Gloves are powder-free, and offer strong barrier protection
Have Questions?
Catalog NumberSize
4472Medium
Catalog number 4472
Price (SGD)
46.00
CS
Add to cart
Size:
Medium
Price (SGD)
46.00
CS
Add to cart
This non-sterile glove is made from a soft, synthetic formulation to offer a comfortable fit and feel. It also has an excellent resistance from a broad range of commonly used chemicals. High tear resistance ensures optimal protection, and a chlorinated inner coating facilitates ease of donning.
  • Excellent resistance to a broad range of commonly used chemicals
  • Protects from Type I Latex allergies
  • Tested for use with chemotherapy drugs
  • See product packaging or contact Customer Service for specific chemotherapy drug permeation times and recommendations
Specifications
ColorBlue
MaterialNitrile
SizeMedium
Glove Cuff StyleBeaded
Unit SizeCS