Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Actual product may vary
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
Have Questions?
Catalog NumberColorProduct Type
4471RedScrew Cap Insert
Catalog number 4471
Price (SGD)
46.00
CS
Add to cart
Color:
Red
Product Type:
Screw Cap Insert
Price (SGD)
46.00
CS
Add to cart
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Highest Quality
  • All caps and inserts are supplied free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Superior Cap Design
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -176°C
Cap Options Simplify Sample Identification
  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors
Specifications
Product TypeScrew Cap Insert
MaterialPlastic
ColorRed
PackagingBulk, 500 Caps
Product LineMatrix™
Barcode2D
SterilitySterile
Capacity (Metric)500 μL and 1.0 mL tubes
For Use With (Equipment)500 μL and 1.0 mL Screwtop Tube Caps
Fit for PurposeSterile and PCR Ready
Unit SizeCS