Search Fisher Scientific
| Catalog Number | Volume (Metric) |
|---|---|
| 4421140 | 1.0 to 5.0 mL |
| 4421130 | 0.4 to 2.0 mL |
| 4421150 | 2.0 to 10.0 mL |
| 4421160 | 5.0 to 30.0 mL |
| 4421170 | 10.0 to 60.0 mL |
Features:
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.
Warranty: 1 year