Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Actual product may vary
Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Have Questions?
Catalog NumberColorSterilityProduct Type
430082ClearNon-sterileStrip Cap
Catalog number 430082
Price (SGD)
292.00
CS
Add to cart
Color:
Clear
Sterility:
Non-sterile
Product Type:
Strip Cap
Price (SGD)
292.00
CS
Add to cart
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
Specifications
FormatStrip Plate
Array1 x 8
Packaging180 Cases, 60 Pk.
ColorClear
Product TypeStrip Cap
MaterialPolyethylene
No. of Wells8
SterilityNon-sterile
No. per Case180
No. per Pack60
Plate CompatibilityImmuno Standard Modules
Quantity80/Cs.
Unit SizeCS
Frequently asked questions (FAQs)
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?
Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.