Search Thermo Fisher Scientific
Avoid breakage and corrosion with these heavy-duty funnels, the toughest large funnels available.
Catalog Number | Diameter (Metric)Top | Capacity (Metric) |
---|---|---|
4260-0060 also known as 4260-0060A | 150 mm | 777 mL |
FIS10-371AA | 101mm | 254 mL |
FIS10-371BB | 150 mm | 777 mL |
FIS10-371CC | 252 mm | 3684 mL |
Avoid breakage and corrosion with Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels, the toughest large funnels available. Lighter weight than glass or metal funnels, these low density polyethylene funnels provide you with long-term use and are a valuble asset to lab safety programs.
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Low Density Polyethylene (LDPE) labware at end of product life. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.