Eco Safety Sign NOTICE Empty Cylinders
Eco Safety Sign NOTICE Empty Cylinders
Actual product may vary
Eco Safety Sign NOTICE Empty Cylinders
The sign is available in 2 sizes, 7 in. x 10 in., and 10 in. x 14 in., and three eco-friendly formats.
Have Questions?
Catalog NumberDimensions (L x W)MaterialMounting Type
4252-010010 x 14 in.PlasticCorner Holes
Catalog number 4252-0100
Price (SGD)
194.00
CS
Add to cart
Dimensions (L x W):
10 x 14 in.
Material:
Plastic
Mounting Type:
Corner Holes
Price (SGD)
194.00
CS
Add to cart
  • Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks. The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
  • Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting
  • Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure
  • All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Specifications
MaterialPlastic
Quantity1 Ea.
Diameter (English)14 in.
Mounting TypeCorner Holes
For Use With (Application)Communication
Height (English)10 in.
Width (English)14 in.
LanguagesEnglish
Dimensions (L x W)10 x 14 in.
Displayed TextNOTICE Empty Cylinders