Search Fisher Scientific
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
| Catalog Number | Diameter (Metric)Top | Capacity (Metric) |
|---|---|---|
| 4250-0100 | 98 mm | 254 mL |
| FIS10-347C | 35 mm | 10 mL |
| FIS10-347B | 54 mm | 41 mL |
| FKS10490143 | 63 mm | 64 mL |
| FIS10-347E | 76 mm | 114 mL |
| FKS10294491 | 76 mm | 114 mL |
| FIS10-347G | 98 mm | 254 mL |
| FIS10-347H | 158 mm | 976 mL |
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.