Matrix™ DeepWell™ Storage Blocks
Matrix™ DeepWell™ Storage Blocks
Actual product may vary
Matrix™ DeepWell™ Storage Blocks
Use these DeepWell™ storage blocks as mother plates when creating compound libraries from automated liquid handling instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityWell ShapeVolume (Metric) Well
4212-11SterileRound1200 μL
4211-11Non-sterileRound1200 μL
Catalog number 4212-11
Price (SGD)
409.00
CS
Add to cart
Sterility:
Sterile
Well Shape:
Round
Volume (Metric) Well:
1200 μL
Price (SGD)
409.00
CS
Add to cart
When creating compound libraries using automated liquid handling instruments, use Thermo Scientific™ Matrix™ DeepWell™ Storage Blocks as a mother plate. These strong blocks are constructed of polypropylene and are chemically resistant, meeting all standard microplate specifications. The unique chimney well design of the 1mL blcok reduces the risk of cross-contamination between wells during pipetting, while the 2mL block has cut-away sides for easy handling and placement on robotic systems.
Warranty: 90 days
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
FormatStandard
Array8 x 12
AutoclavableAutoclavable
Packaging40 Pk., 40 Cases
Binding TypeLow binding PP
Product LineNunc
ColorNatural
Product TypePolypropylene Storage & Sample Prep Plate
For Use With (Equipment)ALPS Heat Sealers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well1200 μL
No. of Wells96
Volume (Metric) Working1000 μL
SterilitySterile
Surface TreatmentNon-treated
Well ShapeRound
No. per Case40
No. per Pack40
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
Unit SizeCS