Matrix™ Blank and Alphanumeric Storage Tubes
Matrix™ Blank and Alphanumeric Storage Tubes
Actual product may vary
Matrix™ Blank and Alphanumeric Storage Tubes
Matrix™ Blank and Alphanumeric Storage Tubes
Matrix™ Blank and Alphanumeric Storage Tubes
Matrix™ Blank and Alphanumeric Storage Tubes
Store valuable samples where 2D tracking is not required with high-density blank and alphanumeric storage tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypePackagingSterilityFit for Purpose
4140
also known as 4140MX
Blank Storage Tube96 Tips/Rack, 8 Racks/Pack, 96 Tips/Rack, 8 Racks/PackNon-sterile, Non-sterilePCR Ready
FKS10785204Blank Storage Tube10 Refill Carriers of 96 TubesSterileSterile and PCR Ready
4247
also known as AZS4247
Blank Storage Tube10 Latch Racks of 96 TubesNon-sterilePCR Ready
Catalog number 4140
also known as 4140MX
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Blank Storage Tube
Packaging:
96 Tips/Rack, 8 Racks/Pack, 96 Tips/Rack, 8 Racks/Pack
Sterility:
Non-sterile, Non-sterile
Fit for Purpose:
PCR Ready
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or DeepWell™ block sample storage.
Highest Quality
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Superior Storage Rack Design
  • Unlike traditional storage tubes or blocks, Thermo Scientific Matrix blank and alphanumeric storage tubes are available in our specially designed, stackable, microplate-footprint Latch Racks of 96 tubes for individual sample, access and removal
  • The Latch Rack design enables multichannel pipette access and eliminates contamination risk with a lid that never touches the bench top when open
  • The Latch Rack is also compatible with automated liquid handling, as the lid can be lifted robotically for high-throughput applications
Clear Designations
  • Matrix alphanumeric storage tubes are clearly marked with high contrast printing on the base of each tube, designating their appropriate well location within a rack, for easy identification
  • Enhance ease of visual identification with colored SepraSeal capping options or labelling on the side of the tube using the ID Scribe instrument
Capping Options
  • Several sealing options are available, including the Thermo Scientific Matrix line of CapMats and CapStrips for sealing multiple tubes and the Thermo Scientific Matrix SepraSeal and DuraSeal capping systems for individual tube sealing, access and piercing
  • Applied with the Thermo Scientific SuperSealer, sealing is secure and consistent every time
Compatible with:
Capping systems, SepraSeals, Duraseals, SuperSealer, ID Scribe
Recommended for:
Individual sample retrieval; shipping samples; storage of DNA, compounds, or any other samples
Specifications
Product TypeBlank Storage Tube
Volume (Metric)100 to 1000 μL
FilteredFiltered
SterilityNon-sterile, Non-sterile
ColorNatural
Capacity (Metric)1.4 mL
MaterialHDPE, HDPE
Clean ClaimsNonpyrogenic, Human DNA-free, RNase-/DNase-free, Sterility Assurance Level 10-3
For Use With (Application)Used for quantitative assays in which sample binding can significantly alter results
Packaging96 Tips/Rack, 8 Racks/Pack, 96 Tips/Rack, 8 Racks/Pack
FormatHinged Rack
Tip StyleRound
Product LineMatrix™
GraduatedYes
Product LineDeckWorks
Capacity (English)0.0473 fl oz.
Dnase FreeYes
Fit for PurposePCR Ready
Unit SizeCS