DeckWorks™ Reload Systems Starter Kits
DeckWorks™ Reload Systems Starter Kits
Actual product may vary
DeckWorks™ Reload Systems Starter Kits
Contains one DeckWorks Reload System with standard tips and empty hinged racks
Have Questions?
Catalog NumberVolume (Metric)Color
4131
also known as CD413X1A
1 to 200 μLNatural
Catalog number 4131
also known as CD413X1A
Price (SGD)
19,337.00
EA
Add to cart
Volume (Metric):
1 to 200 μL
Color:
Natural
Price (SGD)
19,337.00
EA
Add to cart
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
  • 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
  • 8 empty racks accompany 1000 μL volume tips.
Specifications
Volume (Metric)1 to 200 μL
SterilityNon-sterile
ColorNatural
For Use With (Application)Designed to allow to maximize storage space and help reduce the production and waste of plastic materials
FormatRefill
Tip StyleRound
PackagingStarter kit
GraduatedYes
Product LineDeckWorks