Capit-All™ Screw Cap Tube Capper/Decapper
Actual product may vary
Capit-All™ Screw Cap Tube Capper/Decapper
Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily.
Have Questions?
Catalog NumberDecapping FormatFor Use With (Equipment)Speed
4111MAT96-ChannelMatrix Screw Top Caps∼10 sec./Rack
Catalog number 4111MAT
Price (SGD)
128,763.00
Special offer
Ends: 30-Jun-2026
143,070.00
Save 14,307.00 (10%)
Each
Decapping Format:
96-Channel
For Use With (Equipment):
Matrix Screw Top Caps
Speed:
∼10 sec./Rack
Price (SGD)
128,763.00
Special offer
Ends: 30-Jun-2026
143,070.00
Save 14,307.00 (10%)
Each
Maintain sample integrity with a hands-free method of capping and decapping tubes quickly and easily with the Thermo Scientific™ Capit-All™ Screw Cap Tube Capper/Decapper. Designed for benchtop or integrated use, Capit-All instruments cap and decap 24-, 48-, and 96- format Thermo Scientific™ Matrix™ and Thermo Scientific™ Nunc™ ScrewTop tubes with precision and ease. All instruments cap tubes to optimal torque.
Enhanced Productivity
  • Increase throughput by capping and decapping an entire rack at a time in approximately 10 seconds
  • Automatically remove and replace caps on tubes of differing heights, even in the same rack, as well as tubes not properly seated in racks
  • Completely replace manual capping and decapping methods
Superior Sample Protection
  • Caps all tubes in a rack to the optimal torque level, maintaining sample integrity and preventing sample loss during storage
  • Safety features ensure racks are loaded in the correct orientation and that caps are not cross-threaded or overtightened
  • Drip tray prevents cross contamination of samples during addition and removal of tube racks
  • Internal construction ensures debris is not introduced into uncapped tubes
Ease of Use
  • The simple, four-button user interface enables laboratory personnel of any level to operate the instrument
  • Easily integrated into automated systems
  • Easily maintained and able to process up to a million tubes before requiring preventative maintenance
Specifications
Compatible Storage TubesMatrix Style Screw Top Capped Tubes
Decapping Format96-Channel
DescriptionCapper/Decapper
Dimensions (L x W x H)14.2 x 12.9 x 22 in. (32.8 x 36.0 x 56.0 cm)
Electrical Requirements110 to 240VAC 50/60Hz
Net Weight (English)97 lb.
Net Weight (Metric)45 kg
No. per Case1
No. per Pack1
OperationManual via Push Buttons
Voltage110/240 V
TypeCapper/Decapper
For Use With (Equipment)Matrix Screw Top Caps
Frequency50/60 Hz
Speed∼10 sec./Rack
Unit SizeEach