SuperSealer
SuperSealer
Actual product may vary
SuperSealer
SuperSealer
Thermo Scientific
SuperSealer
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer.
Have Questions?
Catalog NumberProduct Type
4110-11Microplate Sealer
Catalog number 4110-11
Price (SGD)
15,462.00
EA
Add to cart
Product Type:
Microplate Sealer
Price (SGD)
15,462.00
EA
Add to cart
Ensure a proper seal on plates, blocks and storage tube racks with the Thermo Scientific™ SuperSealer. Choose individual sealing options such as Thermo Scientific™ Matrix™ SepraSeal mats for liquid handling and single tube accessibility, or seal entire plates and blocks for storage with Thermo Scientific™ Nunc™ CapMats. Effectively apply adhesive tape seals and ensure a tight seal with the included SuperSealer Adhesive Seal Mat.
Sealing Simplicity
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
Sealing Flexibility
  • Designed to work with Thermo Scientific™ SepraSeal mats, CapMats and, with the SuperSealer AdhesiveSeal Mat, any pre-cut adhesive tape seal, you can be assured a proper seal regardless of sealing option
Includes: Automatic cap, tape and SepraSeal mat applicator
Warranty: 1 year
Specifications
DescriptionThermo Scientific Super Sealer
No. per Pack1 Ea.
Product TypeMicroplate Sealer
Unit SizeEA