Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Have Questions?
Change view
Catalog Number
Type
Voltage
4000665
Thermal Data Printer
220 V
4000565
Thermal Data Printer
120 V
4000566
Thermal Printer Paper
Not Applicable
3 Options
Catalog number 4000665
Price (SGD)
3,140.00
EA
Add to cart
Type:
Thermal Data Printer
Voltage:
220 V
Price (SGD)
3,140.00
EA
Add to cart
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
Model 4000566 - Thermal printer paper (5 rolls/package)
Warranty varies by country, please contact us for details
Specifications
TypeThermal Data Printer
For Use With (Equipment)All CryoMed Controlled-Rate Freezers
WarrantyWarranty varies by country, please contact us for details