CryoMed™ Thermocouple Sensors
CryoMed™ Thermocouple Sensors
Actual product may vary
CryoMed™ Thermocouple Sensors
CryoMed™ Thermocouple Sensors
CryoMed™ Thermocouple Sensors
Sample thermocouple sensors
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
4000393Ribbon Type
4000381Thermocouple Sensor
Catalog number 4000393
Price (SGD)
211.00
EA
Add to cart
Type:
Ribbon Type
Price (SGD)
211.00
EA
Add to cart
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
  • Include male plug
For controlled-rate freezing applications
Specifications
TypeRibbon Type
Plug TypeMale
HoldsCryo Tube, Sheath Straw, Vial
Unit SizeEA