T-valve Assembly for LN2 Supply Tank
T-valve Assembly for LN<sub>2</sub> Supply Tank
Actual product may vary
T-valve Assembly for LN2 Supply Tank
Connect two hoses to a LN2 tank with this T-valve assembly, which includes a manual valve and T-fitting.
Have Questions?
Catalog number 4000290
Price (SGD)
1,191.00
EA
Add to cart
Price (SGD)
1,191.00
EA
Add to cart
Connect two hoses to a LN2 tank with this Thermo Scientific™ T-valve assembly for LN2 supply tank, which includes a manual valve and T-fitting.
Specifications
DescriptionManual valve and T fitting
TypeManual Valve and T-Fitting
For Use With (Equipment)CryoPlus 1,2,3,4
DefrostManual
Unit SizeEA