Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Actual product may vary
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsPositionsFor Use With (Equipment)
398331Boxes (2 in.)16 Boxes/RackUpright Freezers (17.3, 23 cu.ft.)
1950638Microplates24 Plates/RackEXF320, EXF400, HFU320B, HFU400B (5 shelf) (17.3, 23 cu.ft.)
FIS19-506-39Microplates28 Plates/RackRevco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.)
398326Boxes (2 in.)16 Boxes/RackUpright Freezers (17.3, 23 cu.ft.)
398330Boxes (3 in.)12 Boxes/RackUpright Freezers (17.3, 23 cu.ft.)
398327Boxes (2 in.)12 Boxes/RackUpright Freezers (17.3, 23 cu.ft.)
Catalog number 398331
HSN 84189900
Price (SGD)
121.00
Special offer
Ends: 30-Jun-2026
Save 40.00 (25%)
Each
Holds:
Boxes (2 in.)
Positions:
16 Boxes/Rack
For Use With (Equipment):
Upright Freezers (17.3, 23 cu.ft.)
Price (SGD)
121.00
Special offer
Ends: 30-Jun-2026
Save 40.00 (25%)
Each
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country, please contact us for details
Specifications
Description2 in. Box Rack - Sliding Drawer
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions16 Boxes/Rack
For Use With (Equipment)Upright Freezers (17.3, 23 cu.ft.)
Width (English)5.5 in.
Height (English)9.4 in.
Depth (English)22 in.
Depth (Metric)55.9 cm
Product TypeBox Racks - Sliding Drawer
Height (Metric)23.9 cm
Width (Metric)14 cm
Unit SizeEach