CellBIND™ Polystyrene Roller Bottles with Easy Grip Cap
CellBIND™ Polystyrene Roller Bottles with Easy Grip Cap
Actual product may vary
CellBIND™ Polystyrene Roller Bottles with Easy Grip Cap
Treated with patented Corning CellBIND™ surface treatment
Have Questions?
Catalog NumberCell Growth AreaClosure TypeClosure Color
3907850 cm2Easy GripOrange
Catalog number 3907
Price (SGD)
40,061.00
EA
Add to cart
Cell Growth Area:
850 cm2
Closure Type:
Easy Grip
Closure Color:
Orange
Price (SGD)
40,061.00
EA
Add to cart
  • Easy Grip Vent Cap is designed for applications requiring consistent gas exchange and features large knurls designed for ergonomic handling
  • Bottles are manufactured from virgin polystyrene, caps from high density polyethylene
  • Both resins meet the USP Class VI requirements for plastic containers and closures
  • Treated with a Corning™ CellBIND™ surface treatment for enhanced cell attachment
  • One piece seamless construction with printed graduations
  • Printed lot numbers to aid in product traceability
  • Sterilized by gamma radiation and nonpyrogenic
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Diameter (English)4.6 in.
Diameter (English) Outer Neck1.77 in.
Diameter (Metric) Outer Neck45 mm
MaterialPolystyrene
No. per Case40/Cs.
No. per Pack2/Pk.
SterilitySterile
Cell Growth Area850 cm2
Closure ColorOrange
Surface TreatmentCorning CellBIND Surface
GraduatedNo
MouthWide
Height (Metric)271 mm
Height (English)10.66 in.
Closure MaterialHDPE
Closure Size48 mm
Diameter (Metric)117 mm
Closure TypeEasy Grip
Unit SizeEA