Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Actual product may vary
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (Metric)Thread StyleStyle
377267Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
374080Conical (Self-standing)0.5 mLInternalNo Writing Surface, A-R Base
375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-164NConical (Self-standing)1 mLExternalWriting Surface, A-R Base
FKS10544641Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
NNC375353Conical (Self-standing)1 mLExternalWriting Surface, A-R Base
FIS12-565-297Conical (Self-standing)1 mLExternalNo Writing Surface, A-R Base
NNC377224Conical (Self-standing)1 mLInternalWriting Surface, A-R Base
366656Conical (Self-standing)1 mLInternalWriting Surface
375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
NNC375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
FKS10080601Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
NNC347627Round (Self-standing)1.8 mLExternalNo Writing Surface, A-R Base
FIS12-565-167NRound (Self-standing)1.8 mLInternalWriting Surface, A-R Base
FKS11341675Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
368632Round (Self-standing)1.8 mLInternalWriting Surface
FIS12-565-171NRound (Self-standing)1.8 mLInternalWriting Surface
FKS10674511Round (Self-standing)1.8 mLInternalWriting Surface
363401Round1.8 mLInternalWriting Surface
FKS10004220Round1.8 mLInternalWriting Surface
NNC363401Round1.8 mLInternalWriting Surface
351934
also known as 35193 4
Round1.8 mLInternalWriting Surface, No Cap
379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FIS12-565-162NRound (Self-standing)3.6 mLInternalWriting Surface, A-R Base
NNC379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
FKS10789871Round3.6 mLInternalWriting Surface
337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
FIS12-565-291Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
NNC337516Round (Self-standing)4.5 mLExternalWriting Surface, A-R Base
347643Round (Self-standing)4.5 mLExternalNo Writing Surface, A-R Base
FIS12-565-161NConical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
FKS10716922Conical (Self-standing)4.5 mLInternalWriting Surface, A-R Base
363452Round4.5 mLInternalWriting Surface
FIS12-565-173NRound4.5 mLInternalWriting Surface
FKS10358131Round4.5 mLInternalWriting Surface
Catalog number 377267
HSN 39269099
Price (SGD)
435.00
PK
Bottom Shape:
Round (Self-standing)
Capacity (Metric):
1.8 mL
Thread Style:
Internal
Style:
Writing Surface, A-R Base
Price (SGD)
435.00
PK
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Diameter (English) Outer0.492 in.
Capacity (Metric)1.8 mL
Bottom ShapeRound (Self-standing)
MaterialPolypropylene
Thread StyleInternal
Length (English)1.929 in.
Length (Metric)49 mm
Packaging450 Tubes
StyleWriting Surface, A-R Base
RNase FreeYes
Vapor LN2 (to -150°C)Yes
Product LineNunc
SterilitySterile
Anti-RotationYes
Packaging Quantity450 Pk.
Closure MaterialPolypropylene
Capacity (English)0.0608 oz.
Dnase FreeYes
Closure TypeScrew Cap
PrintedYes
Diameter (Metric)12.5 mm
Product TypeCryogenic Storage Tube
Green FeaturesACT Ecolabel (Silver)
Unit SizePK
Frequently asked questions (FAQs)
Will the heat required to properly seal Nunc CryoFlex Tube Wrap adversely affect my cells?
Nunc Biobanking and Cell Culture Cryogenic Tubes are normally placed in a 4 degrees C ice bath during cell inoculation in the cryogenic tube. The heat required to shrink the polyethylene Nunc CryoFlex Tube Wrap will not raise the internal temperature enough to damage the cells. At this stage, cryoprotective agents are added.
What protocol should I follow for thawing cells, if my Nunc Biobanking and Cell Culture Cryogenic Tubes were previously immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap?
We recommend the following protocol for thawing cells immersed in liquid nitrogen without Nunc CryoFlex Tube Wrap:
- Wear appropriate protective clothing including gloves, face guard, etc., during all transfer processes.
- The Nunc Biobanking and Cell Culture Cryogenic Tubes should be stored at least 24 hr in the gas phase of the liquid nitrogen freezer. This should allow trapped liquid nitrogen to evaporate and escape slowly, thereby reducing the possible pressure in the cryogenic tube.
- After at least 24 hr storage in the gas phase, the cryogenic tube may be put into a water bath and covered with a towel.
- After thawing, the contents of the cryogenic tube can be transferred.
Can I immerse Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid nitrogen (liquid phase) for storage?
Of the Nunc Biobanking and Cell Culture Cryogenic Tubes, we recommend the internally threaded design, incorporating a silicon gasket, for liquid nitrogen immersion. At ultra low temperatures, any sealing material becomes inflexible. A theoretical risk of leakage is present when any mechanical closure is used. To facilitate leakproof storage, mechanically closed containers must be hermetically sealed in a protective envelope.
The Nunc cryo products includes Nunc CryoFlex Tube Wrap, a heat sealable polyethylene tubing designed for cryogenic tube encasement. The use of cryogenic tubes for freezing in liquid nitrogen (liquid phase) storage without CryoFlex tubing poses a safety hazard.
WARNING: Do not use Nunc Biobanking and Cell Culture Cryogenic Tubes in liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex Tube Wrap (Cat. No. 343958). Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. Use appropriate safety procedures as outlined in the Nalge Nunc International Cryopreservation Manual when handling and disposing of the vials.
What is the difference between your Nunc Biobanking and Cell Culture Cryogenic Tubes with internal threads and external threads?
Internally threaded Nunc Biobanking and Cell Culture Tubes are designed for vapor phase liquid nitrogen freezing. The cap incorporates a silicon gasket to enhance sealing properties.
Externally threaded Nunc Biobanking and Cell Culture Tubes are designed for mechanical freezing. The external screw cap reduces risk of contamination due to handling.
Can I autoclave Nunc Biobanking and Cell Culture Cryogenic Tubes?
We do not recommend autoclaving Nunc Biobanking and Cell Culture Tubes, even though they are made of polypropylene with polyethylene caps and stoppers.