Nunc™ 2 mL Internally-Threaded Universal Tubes
Actual product may vary
Nunc™ 2 mL Internally-Threaded Universal Tubes
Nunc™ 2 mL Internally-Threaded Universal Tubes
Nunc™ 2 mL Internally-Threaded Universal Tubes
Nunc™ 2 mL Internally-Threaded Universal Tubes
Nunc™ 2 mL Internally-Threaded Universal Tubes
Thermo Scientific
Nunc™ 2 mL Internally-Threaded Universal Tubes
Experience the best features of our storage tube portfolio: universal cap, universal latch rack and free-standing tube design -- all for either benchtop or automation use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeCodingPackaging
374510Universal Tubes in Latch Rack2D Barcode (Tube Only)10 Racks of 48 Tubes
374512Universal Tube2D BarcodeBulk
Catalog number 374510
HSN 39269099
Price (SGD)
861.00
Special offer
Ends: 30-Jun-2026
1,414.00
Save 553.00 (39%)
CS
Type:
Universal Tubes in Latch Rack
Coding:
2D Barcode (Tube Only)
Packaging:
10 Racks of 48 Tubes
Price (SGD)
861.00
Special offer
Ends: 30-Jun-2026
1,414.00
Save 553.00 (39%)
CS
Experience the best features of the Thermo Scientific™ storage tube portfolio: universal cap, universal latch rack and free-standing tube. This versatile, 2.0mL internally threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • Internal threads limit contamination
  • Automation-pickable in universal latch rack
  • Barcoded rack option offers additional tracking assurance
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 10-6 and is made with class VI medical grade resins
Specifications
Chemical Name or MaterialRN 1747
Coding2D Barcode (Tube Only)
CAS1024448-59-6
Vial TypeInternal Thread, Flat Bottom
ColorNatural
Packaging10 Racks of 48 Tubes
Recommended StorageStore at +4°C
For Use With (Equipment)Biobank, Cryostorage
IncludesWhite patch and graduations
Quantity480
Molecular FormulaC17H18ClN3O4S
Quantity10 mg
Product LineNunc
SterilitySterile
Synonym4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
InChI KeyZNLVYSJQUMALEO-UHFFFAOYSA-N
SMILESC1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]
IUPAC Name1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
Molecular Weight (g/mol)395.858
TypeUniversal Tubes in Latch Rack
Closure ColorClear
PubChem CID5068295
Capacity (Metric)2 mL
Formula Weight395.87
MaterialPlastic
Percent Purity>98%
Unit SizeCS
Frequently asked questions (FAQs)
What is the Thermo Fisher alternative to Corning® 2 mL Internal Threaded Polypropylene Cryogenic Vial with 1D and 2D Bar codes, Product Number 8670?
The following alternatives for the Corning Cryogenic Vial, Product Number 8670:
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374510, 374510-BR)
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374512). Please note that this product is FDA registered and CE-marked.
- Nunc 1.8mL Externally-Threaded Universal Tubes and Racks (Cat. No. 374502, 374502NOV
- Matrix ScrewTop Tri-coded Tubes (Cat. No. 3740-WP1D)