Digital Hand-Held Pen Refractometer, PEN-Urine S.G.
Digital Hand-Held Pen Refractometer, PEN-Urine S.G.
Actual product may vary
Digital Hand-Held Pen Refractometer, PEN-Urine S.G.
The Atago™ PEN-Urine S.G. Digital Hand-Held Pen Refractometer uses the refractive index method to measure the specific gravity of urine which varies with concentration.
Have Questions?
Catalog number 3741
Price (SGD)
758.00
CS
Add to cart
Price (SGD)
758.00
CS
Add to cart
  • Measurement time is just 2 seconds
  • Water resistant
Specifications
Scale Range1.0000 to 1.0600 Urine Specific Gravity
ModelPEN-Urine S.G.
Sample VolumeSmall volumes
CalibrationWith water only
Accuracy±0.0010
ResolutionUrine S.G. 0.0001
Measurement Time2 sec.
For Use With (Application)Measuring specific gravity of urine
IP RatingIP65 Water resistant (Prism head: IP67)
Weight (English)0.1 lb.
Weight (Metric)70 g
Height (English)0.7 in.
Height (Metric)18 mm
Width (English)6.2 in.
Width (Metric)160 mm
Ambient Temperature (Metric)10°C to 40°C
Temperature Range (Metric)10°C to 40°C
Power Supply Details1xAAA Alkaline Battery
Unit SizeCS