Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Actual product may vary
Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Nunc™ Non-Coded Cryobank Vial Systems
Take traditional cryostorage to the next level, with standard microplate format racks, accommodating denser storage capability compared to standard cryoboxes.
Have Questions?
Catalog NumberCapacity (Metric)Closure ColorSterilityPackaging
3740891 mLClearSterileRacked, 96 x 10
Catalog number 374089
Price (SGD)
1,010.00
CS
Add to cart
Capacity (Metric):
1 mL
Closure Color:
Clear
Sterility:
Sterile
Packaging:
Racked, 96 x 10
Price (SGD)
1,010.00
CS
Add to cart
Store diagnostic and biological samples down to vapor-phase liquid nitrogen in Thermo Scientific™ Nunc™ Non-Coded Cryobank Vial Systems, compatible with capping/decapping equipment and labeling options. Cryobank products are available in a volume range of 0.5L through 5.0L, and complimented by a range of certifications.
Secure Tracking
  • Solid one-piece construction eliminates the risk of labels falling off during cryogenic storage, preserving the identity of valuable sample material
Superior Storage Format
  • Cryobank tubes are available in space-conscious microplate format racks or can be stored in traditional cryoboxes, accommodating your preferred storage format
  • Microplate format is compatible with your existing multichannel, manual pipettes and automated liquid-handling equipment
  • Unique anti-rotation feature allows compatibility with automated decapping/capping equipment in single, 8-channel, or full-rack options to accommodate multiple throughput requirements
  • TPE-gasketed polypropylene cap provides superior seal during high- or low-temperature storage
  • Low-binding surface allows maximum recovery of low-concentration samples
Certified to meet your storage needs
  • CE certified and conforms to IATA requirements for diagnostic use and transportation
  • Certified DNAse/RNAse free (0.5/1.0mL), non-pyrongenic (LAL-test), non-toxic (USP class VI test)
  • Sterility assurance level 10-6 for the most sensitive sample storage
Stringent Quality Control
  • Leak-testing validation performed during the manufacturing process to ensure sample integrity is maintained
Specifications
Closure ColorClear
Description374089
Capacity (Metric)1 mL
MaterialPolypropylene (Cap)
SterilitySterile
TypeCryobank
CodingNone
Vial TypeCryobank
ColorClear (Closure)
PackagingRacked, 96 x 10
No. per Pack960
No. per Case960
Product LineNunc™
Unit SizeCS