PTFE-Coated Micro Double Spatulas
PTFE-Coated Micro Double Spatulas
Actual product may vary
PTFE-Coated Micro Double Spatulas
Well-suited for small quantity applications and narrow-format tubes, these double-ended, PTFE-coated spatulas reduce material sticking. Bochem™ PTFE-Coated Micro Double Spatulas are also gentle on delicate, scratchable surfaces.
Have Questions?
Catalog NumberLength (Metric)Length (Metric) BladeDiameter (Metric) OuterWidth (Metric)
3711
also known as QSF3711A
185 mm50 mm2 mm4 mm
Catalog number 3711
also known as QSF3711A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
185 mm
Length (Metric) Blade:
50 mm
Diameter (Metric) Outer:
2 mm
Width (Metric):
4 mm
  • Non-stick PTFE coating.
  • Double-ended for versatility.
  • Slim ends for easy container access.
  • One rounded end and one flat end.
Specifications
DescriptionDouble Ended
CoatingPTFE
Length (Metric) Blade50 mm
Width (Metric)4 mm
Width (Metric) Blade2 mm
Diameter (Metric) Outer2 mm
Length (Metric)185 mm
Unit SizeCS