FirmGrip Shoe Covers
FirmGrip Shoe Covers
Actual product may vary
FirmGrip Shoe Covers
FirmGrip Shoe Covers
FirmGrip Shoe Covers
FirmGrip Shoe Covers
FirmGrip Shoe Covers
Shoe covers provide an extra sticky firm and secure grip to reduce the risk for slips, trips, and falls.
Have Questions?
Catalog NumberSizeMaterialColor
3701LargeProprietary Laminated Non-woven FabricWhite
Catalog number 3701
Price (SGD)
138.00
PK
Add to cart
Size:
Large
Material:
Proprietary Laminated Non-woven Fabric
Color:
White
Price (SGD)
138.00
PK
Add to cart
FirmGrip non-skid shoe covers are designed for tough working environments. Not only do they protect from particulates, but they also safeguard against light liquid splashes. Using a multi-layer fabric and textured finish, these durable shoe covers offer the traction to get things done without slipping. A strong elastic closure keeps shoe covers tightly in place
  • Textured surface for a firm grip in slick or wet environments
  • Strong elastic closure provides a secure fit
  • Size Large fits men's sizes 6 to 11
  • Size Extra Large fits men's sizes 12 to 16
  • Common applications include: Healthcare, Construction, Realtors, Movers, Delivery Services, Food Processing, Cleanrooms, Installers, and more
  • Can be worn in indoor and outdoor environments
Specifications
SizeLarge
MaterialProprietary Laminated Non-woven Fabric
Latex FreeYes
ColorWhite
DisposableSingle-use
SterilityNon-sterile
Closure TypeElastic Ankle
Sole TypeNon-skid
For Use With (Application)Construction, Realtors, Movers, Delivery Services, Food Processing, Cleanrooms, Installers
Packaging Quantity300/Cs.
ResistanceDry Particulates, Splash
Product TypeControlled Environments Shoe Covers
Unit SizePK