Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Actual product may vary
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Thermo Scientific
Nalgene™ Polypropylene Economy Plastic Graduated Cylinders
Dispense liquids easily with simple to read, molded-in graduations that won't rub off using these affordably-priced graduated cylinders.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
3664-0100100 mL1 mL
FIS08-572-6A10 mL0.2 mL
3664-005050 mL1 mL
3664-0250250 mL2 mL
FIS08-572-6E250 mL2 mL
3664-0500500 mL5 mL
FIS08-572-6F500 mL5 mL
FIS08-572-6G1000 mL10 mL
Catalog number 3664-0100
HSN 39269099
Price (SGD)
10.00
EA
Add to cart
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (SGD)
10.00
EA
Add to cart
Dispense liquids easily with simple to read, molded-in graduations that won't rub off. Affordably priced Thermo Scientific™ Nalgene™ Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
  • Translucent
  • Non-wetting
  • Economical and recyclable; designed for single-use
  • No meniscus to confuse readings, allowing for accurate measurements
  • Temperature resistance up to 135°C (275°F)
  • Large, ribbed, rounded base that provides reinforcement and stability, preventing rolling or tipping
  • Calibrated “To Contain“ and “To Deliver” at 20°C
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.
Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material.  Packaging bags are made from LDPE and are readily recyclable once the label is removed.   Please continue to cycle and recycle your cartons and bags.
.
Specifications
Capacity (Metric)100 mL
MaterialPolypropylene
Item DescriptionGraduated Cylinder Economy PP 100mL
No. per Pack1
No. per Case12
CalibrationTo Contain
Diameter (English)1.11 in.
Diameter (Metric)28.2 mm
Graduations1 mL
Max. Temperature120°C
ShapeRound
Diameter (Metric) Inside39.4 mm
Diameter (English) Inside1.55 in.
Max. Temperature (English)248°F
For Use With (Application)Laboratories
Base TypeRound
Height (English)9 in.
Height (Metric)228.6 mm
TypeGraduated Cylinder
ACT LabelYes
Unit SizeEA
Frequently asked questions (FAQs)
Do you offer class A plastic graduated cylinders?
We do not offer plastic graduated cylinders with Class A accuracy. It is very difficult to reach Class A accuracy with plastic, which is why they are almost always glass.
What is the difference between your Class B and Economy graduated cylinders?
Class B cylinders are certified to meet ASTM accuracy and precision standards. Economy cylinders bear no specific accuracy or precision certification. It is important to note that after autoclaving, even certified products may no longer be as accurate.