Hi-Temp Vacuum Ovens
Hi-Temp Vacuum Ovens
Actual product may vary
Hi-Temp Vacuum Ovens
Ideal for higher temperature drying under vacuum, this vacuum ovens line provides temperatures up to 280°C, with peace of mind due to built-in safety controller and overtemperature protection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltageWattageCapacity (Metric)
3628A-1240 V1400 w42.4 L
3625A-1240 V1700 w18.4 L
Catalog number 3628A-1
Price (SGD)
18,149.00
EA
Add to cart
Voltage:
240 V
Wattage:
1400 w
Capacity (Metric):
42.4 L
Price (SGD)
18,149.00
EA
Add to cart
Ideal for higher temperature drying under vacuum, this vacuum ovens line provides temperatures up to 280°C, with peace of mind due to built-in safety controller and overtemperature protection.
Features include:
  • Two convenient sizes: Small and large model, 18L (0.65 cu.ft.) and 42.4L (1.5 cu.ft.)
  • Temperature range: Ambient +10°C / 50°C to 280°C
  • Radiant warm wall heaters mounted on the outside conserve chamber working space and help eliminate potential hazards of open wire heaters
  • Compression fittings on the side of the unit accept 6.4mm (0.25 in.) outer diameter hard tubing for pump and air connections
  • Vacuum and air lines of corrosion-resistant stainless-steel tubing optimize chamber cleanliness and long-term performance
  • Durable exterior is powder-coated heavy-gauge steel
  • Glass window in the door allows for sample viewing
  • Two adjustable chrome-plated shelves help improve heat conduction or transfer to samples; easily removable for cleaning
Small models 3625A and 3625A-1
  • Dual-electronic control system with dial thermometer (no display)
  • For operational convenience, all controls and indicators are located on the front panel: power switch and three indicators, purge gas and vacuum control valves, a 0-to-30-inch vacuum gauge and a temperature control
Large models 3628A and 3628A-1
  • Single set point controller
  • Seven-day programmable timer that cycles oven on or off up to eight times per day with automatic temperature/ vacuum control in real time steps
  • Dual displays provide constant day/ time and temperature/pressure updates every 5 seconds
Recommended for:
  • Desiccation
  • Vacuum embedding
  • Plating
  • Process control
  • Inert atmosphere
Specifications
Capacity (English)1.5 cu.ft.
Chamber MaterialStainless Steel
Description240V 1400w 6A; 1.5 cu. ft. (42.4L)
Capacity (Metric)42.4 L
Temperature (Metric)50°C to 280°C
TypeVacuum Oven
DisplayLED (2)
MaterialPowder-coated Heavy-gauge Steel
No. of Shelves2
Shelf MaterialChrome-plated
Temperature Uniformity±5°C at 100°C 25 ft.Hg
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 30 x 30 cm)
Width (English)23 in.
Width (English) Interior12 in.
Width (Metric)58 cm
Width (Metric) Interior30 cm
Height (English)22 in.
Height (English) Interior12 in.
Height (Metric)46 cm
Height (Metric) Interior30 cm
Depth (English)26 in.
Depth (English) Interior18 in.
Depth (Metric)66 cm
Depth (Metric) Interior46 cm
Shipping Weight (English)205 lb.
Shipping Weight (Metric)92.9 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Wattage1400 w
Amperage6 A