ART™ Non-Filtered Gel 200 Universal-Fit Pipette Tips
ART™ Non-Filtered Gel 200 Universal-Fit Pipette Tips
Actual product may vary
ART™ Non-Filtered Gel 200 Universal-Fit Pipette Tips
For loading specific types of gels
Have Questions?
Catalog NumberFormatTip StyleMaterial
3621
also known as P3621
RackGel Loading, RoundPolyacrylamide, Polyacrylamide
Catalog number 3621
also known as P3621
Price (SGD)
75.00
PK
Add to cart
Format:
Rack
Tip Style:
Gel Loading, Round
Material:
Polyacrylamide, Polyacrylamide
Price (SGD)
75.00
PK
Add to cart
For loading specific types of gels
Loading acrylamide or even agarose gels with standard pipet tips is an often time-consuming and frustrating process. These gel-loading tips were designed specifically for this application, for optimal results every time. Length: 83mm; Grad: 5μL.
Round Gel 200 (3621):
  • For loading gels 0.6mm thick and thicker
  • For agarose, 2-D, SDS-PAGE and protein electrophoresis
Flat Gel 200:
  • For loading gels up to 0.4mm thick
  • For polyacrylamide and DNA sequencing gels
Specifications
Cap StyleSnap Cap
Certifications/ComplianceRNase, DNase-free
Volume (Metric)200 μL
Closure ColorNatural
Closure MaterialPolypropylene
FilteredNon-filtered
SterilityNon-sterile, Non-sterile
AutoclavableAutoclavable
Capacity (Metric)1.7 mL
ColorNatural
MaterialPolyacrylamide, Polyacrylamide
Thickness0.6mm
For Use With (Application)For loading specific types of gels
No. per Pack500/Pk.
Packaging5000/Pk.
FormatRack
Tip StyleGel Loading, Round
GraduatedYes
Product LineART
Closure TypeSnap Cap
Graduations5μL
Bottom ShapeConical
Max. RCF17,000 x g
Product TypeMicrocentrifuge Tube
Unit SizePK