MaxQ™ 3000/5000 Dedicated Platforms with Clamps
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
Actual product may vary
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
MaxQ™ 3000/5000 Dedicated Platforms with Clamps
Use these dedicated platforms with Thermo Scientific™ MaxQ™ 3000 and 5000 Shakers.
Have Questions?
Catalog NumberProduct TypeFor Use With (Equipment)Length (English) PlatformNo. of Positions
3599-63Adjustable Vessel PlatformMaxQ™ 3000 and MaxQ™ 5000 Shakers Only30 in.Adjustable
Catalog number 3599-63
Price (SGD)
9,445.00
EA
Add to cart
Product Type:
Adjustable Vessel Platform
For Use With (Equipment):
MaxQ™ 3000 and MaxQ™ 5000 Shakers Only
Length (English) Platform:
30 in.
No. of Positions:
Adjustable
Price (SGD)
9,445.00
EA
Add to cart
Achieve maximum capacity if only using a single vessel size such as a flask with Thermo Scientific™ MaxQ™ 3000/5000 Dedicated Platforms with Clamps. Use these dedicated platforms with MaxQ™ 3000 and 5000 Shakers.
  • Clamps come installed to provide maximum capacity
Specifications
Product TypeAdjustable Vessel Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)30 x 18 in.
Length (English) Platform30 in.
Length (Metric) Platform76 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
DescriptionAdjustable vessel platform
For Use With (Equipment)MaxQ™ 3000 and MaxQ™ 5000 Shakers Only
No. of PositionsAdjustable
Unit SizeEA