Check Order Status
Reynold's Wrap Aluminum Foil
Actual product may vary
Reynold's Wrap Aluminum Foil
Pure aluminum foil
Have Questions?
Catalog Number
Length (English)
Width (English)
3591
also known as OM3591C
75 ft.
12 in.
Catalog number
3591
also known as OM3591C
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
75 ft.
Width (English):
12 in.
Sheets are ideal for covering flasks, beakers, petri dishes, weigh boats, disposable crucibles, and wrapping samples.
Specifications
Applications
Ideal for covering flasks, beakers, petri dishes, weigh boats, disposable crucibles, and wrapping samples
Length (English)
75 ft.
Product Type
Aluminum Foil
Width (English)
12 in.
Material
Aluminum