Specifications
Chemical Name or MaterialSodium meta-Bisulfite
Volume (Metric)200 μL
CAS7681-57-4
FilteredNon-filtered
SterilitySterile
ColorNatural
Quantity2.5 kg
Length (English)2 in.
Length (Metric)5.07 cm
Clean ClaimsDNase, RNase, DNA, ATP and Endotoxin Free
FormatRack
Molecular FormulaNa2O5S2
MDL NumberMFCD00167602
Synonymsodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
For Use With (Equipment)Universal Fit
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular Weight (g/mol)190.09
Product LineART
PubChem CID656671
Formula Weight190.11
Fit for PurposeSterile
GradeMulti-Compendial
PackagingPoly Bottle
Specific Gravity1L = 1.48kg
Unit SizePack