MaxQ™ Shaker Universal Platforms
MaxQ™ Shaker Universal Platforms
Actual product may vary
MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Dimensions (L x W)
3522-3Adapter set for stacking two platformsMaxQ™ 30100 and MaxQ™ 30110 Platform13 x 11 in. (33 x 28 cm)
30110DBQUniversal Platform without Clamps, 18 x 18 in.MaxQ™ 2000, 2506, 2508, 4000, 6000 Shaker18 x 18 in. (45.7 x 45.7 cm)
30115BIUniversal Platform without Clamps, 18 x 30 in.MaxQ 3000, 500030 x 18 in. (76 x 45.7 cm)
30128Dual Stacked Universal Platform without Clamps, 18 x 18 in.MaxQ™ 2000, MaxQ™ 6000 Shaker18 x 18 in. (45.7 x 45.7 cm)
Catalog number 3522-3
Price (SGD)
399.00
EA
Add to cart
Description:
Adapter set for stacking two platforms
For Use With (Equipment):
MaxQ™ 30100 and MaxQ™ 30110 Platform
Dimensions (L x W):
13 x 11 in. (33 x 28 cm)
Price (SGD)
399.00
EA
Add to cart
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Product TypeStacking Adapter
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)13 x 11 in. (33 x 28 cm)
Length (English) Platform11 in. (MaxQ 30100 Platforms), 18 in. (MaxQ 30110 Platforms)
Length (Metric) Platform279.4 mm (MaxQ 30100 Platforms), 457.2 mm (MaxQ 30110 Platforms)
DescriptionAdapter set for stacking two platforms
For Use With (Equipment)MaxQ™ 30100 and MaxQ™ 30110 Platform
Unit SizeEA