Check Order Status
Sodium Bicarbonate, Powder, Multi-Compendial, U.S.P., J.T. Baker™
Element Search
Structure Search
Sodium Bicarbonate, Powder, Multi-Compendial, U.S.P., J.T. Baker™
Have Questions?
Catalog Number
Quantity
Packaging
3510-05
2.5 kg
Wide Mouth Amber Glass Bottle
Catalog number
3510-05
Price (SGD)
56.00
PK
Add to cart
Quantity:
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Request bulk or custom format
Price (SGD)
56.00
PK
Add to cart
Specifications
Chemical Name or Material
Sodium Bicarbonate
CAS
144-55-8
Physical Form
Powder
Quantity
2.5 kg
Molecular Formula
CHNaO3
View more