Check Order Status
Marigold™ Foodsure Tear-Resistant Natural Rubber Gloves
Actual product may vary
Marigold™ Foodsure Tear-Resistant Natural Rubber Gloves
Designed with good flexibility, comfort and dexterity
Have Questions?
Catalog Number
Size
3492
7.5
Catalog number
3492
Price (SGD)
48.00
PK
Add to cart
Size:
7.5
Price (SGD)
48.00
PK
Add to cart
Industrial protective glove
Strong yet sensitive formulation
Unsupported flocked natural rubber glove
100% cotton flock for good comfort
Negative pattern grip finish on palm and fingers for good dry grip
Good tear resistance
Tear resistant beaded cuff for easy donning
Not recommended for fatty food
Applications:
Food preparation, processing and handling
Meat and poultry processing
Fish processing
Specifications
Certifications/Compliance
2010 , AQL 1.5
Color
Blue
Latex Free
No
Material
Latex
Size
7.5
Thickness (Metric)
0.43 mm