Check Order Status
18/10 Stainless Steel Chemical Spoon
Actual product may vary
18/10 Stainless Steel Chemical Spoon
Stainless steel spoon for scooping chemicals.
Have Questions?
Catalog Number
Length (Metric)
Length (Metric) Blade
Diameter (Metric) Outer
3465
300 mm
48 mm
5 mm
Catalog number
3465
Price (SGD)
141.00
PK
Add to cart
Length (Metric):
300 mm
Length (Metric) Blade:
48 mm
Diameter (Metric) Outer:
5 mm
Price (SGD)
141.00
PK
Add to cart
Chemical spoon
With spatula handle
Specifications
Quantity
1 Ea.
Product Type
Chemical Scoop, Double
Length (Metric) Blade
48 mm
Width (Metric)
35 mm
Diameter (Metric) Outer
5 mm
Material
Stainless Steel (18/10)
Length (Metric)
300 mm