Digital Urine Specific Gravity Refractometer, UG-α
Digital Urine Specific Gravity Refractometer, UG-α
Actual product may vary
Digital Urine Specific Gravity Refractometer, UG-α
The ATAGO™ Digital Urine Specific Gravity Refractometer UG-α is an upgraded version of previous model UG-1. The minimum scale indication has been improved to display up to the fourth decimal.
Have Questions?
Catalog number 3464
Price (SGD)
946.00
CS
Add to cart
Price (SGD)
946.00
CS
Add to cart
  • Using the principles of light refraction, ATAGO Clinical Refractometers use special scales for clinical purposes such as specific gravity of urine
  • After pressing the START key, with the sample placed on the prism, the measurement values will be displayed on the LCD streaming as an electric message board
Specifications
TypeRefractometer
Refractive Index1.0000 to 1.0600
ModelUG-α
Accuracy± 0.0010
ResolutionMin. Indication: 0.0001
Scale TypeUrine Specific Gravity
Ambient Temperature (Metric)10°C to 35°C
Unit SizeCS