Graduated Safelock Microcentrifuge Tubes
Graduated Safelock Microcentrifuge Tubes
Actual product may vary
Graduated Safelock Microcentrifuge Tubes
Graduated Safelock Microcentrifuge Tubes
Graduated Safelock Microcentrifuge Tubes
Graduated Safelock Microcentrifuge Tubes
Easily confirm your microcentrifuge tube is locked for superior security during centrifugation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityCapacity (Metric)Max. RCFPackaging
3457Sterile1.5 mL24,000 x g50 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
FKS12388202Sterile0.6 mL30,000 x g100 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
FKS11509924Sterile2 mL25,000 x g50 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
Catalog number 3457
Price (SGD)
38.00
PK
Add to cart
Sterility:
Sterile
Capacity (Metric):
1.5 mL
Max. RCF:
24,000 x g
Packaging:
50 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
Price (SGD)
38.00
PK
Add to cart
Find the security of a screw-cap tube with the ease-of-use of a snap-cap tube with Thermo Scientific™ Graduated Safelock Microcentrifuge Tubes. These tubes provide superior security during centrifugation. Simply position the hinged locking ball into the groove to prevent accidental cap opening, avoiding evaporation and ensuring sample integrity.
Safelock Lid Design
  • Simple one-hand operation snaps the hinged cap closed and slides the locking ball into place safeguarding valuable samples
  • Hinged locking ball reduces stress on the thumb
  • Smooth locking surface reduces the possibility of ripped or torn gloves
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Highest Quality
  • Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity and security during boiling, freezing, incubation and centrifugation
  • Exceptionally stable performance under the stress of centrifugation, preventing tube breakage
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Packaging  Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements, individually wrapped sterile tubes are available
Recommended For:
Daily and rigorous laboratory usage including centrifugation, boiling, incubation, freezing or sample storage
Specifications
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Capacity (Metric)1.5 mL
ColorNatural
MaterialPolypropylene
Packaging50 Tubes/Bag, 10 Bags/Unit, 10 Units/Cs.
SterilitySterile
GraduatedYes
Max. RCF24,000 x g
Unit SizePK