18/10 Stainless Steel Section Lifters, with 18/10 Stainless Steel Handle
18/10 Stainless Steel Section Lifters, with 18/10 Stainless Steel Handle
Actual product may vary
18/10 Stainless Steel Section Lifters, with 18/10 Stainless Steel Handle
Manufactured from 18/10 stainless steel
Have Questions?
Catalog NumberDimensions (L x W)
3446
also known as 3446ECONO
150 x 15 mm
Catalog number 3446
also known as 3446ECONO
Price (SGD)
68.00
EA
Add to cart
Dimensions (L x W):
150 x 15 mm
Price (SGD)
68.00
EA
Add to cart
Specifications
Quantity1 Ea.
Product TypeSection Lifter
Length (Metric) Blade50 mm
Width (Metric)15 mm
Blade MaterialStainless Steel (18/10)
Diameter (Metric) Outer0.8mm
Dimensions (L x W)150 x 15 mm
Handle MaterialStainless Steel (18/10)
MaterialStainless Steel (18/10)
Length (Metric)150 mm
Unit SizeEA