Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Actual product may vary
Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Nalgene™ HDPE Tamper Evident Closures, Sterile, Bulk-Pack
Nalgene™ HDPE Tamper Evident Closures are specially designed for use with PET/PETG square media bottles. Featuring a tamper evident break ring design which combines the unique leakproof† nature of the Nalgene bottles/closure system with the possibility to ensure proof of content integrity.
Have Questions?
Catalog NumberThread Size (Metric)
342141-038442 mm
Catalog number 342141-0384
Price (SGD)
216.00
CS
Add to cart
Thread Size (Metric):
42 mm
Price (SGD)
216.00
CS
Add to cart
Nalgene™ HDPE Tamper Evident Closures are specially designed for use with PET/PETG square media bottles.
Features:
  • Available in 38-430 finish
  • Natural HDPE closures are Animal Derived Component (ADC) Free
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish (except 125mL PETG Media Bottles)
  • Leakproof Guarantee when used with Nalgene PET and PETG Media Bottles
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 10-6 Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labeling
Ordering Alerts:
Closures are sterile and bulk packed. Order Nalgene PET or PETG Media Bottles with 38-430 neck finish without closures separately. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
ColorNatural
DescriptionHDPE Tamper Evident Closure
MaterialHDPE
No. per Case240
No. per Pack240
PackagingBulk
SterilitySterile
Thread Size (Metric)42 mm
Neck Finish38-430
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements
Fit for PurposeSterile
Product TypeTamper-evident Closure
For Use With (Equipment)Nalgene PET/PETG square media bottles (except 125mL PETG Media Bottles)
Unit SizeCS