Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Size |
|---|---|---|
| 342089-0032 | 1000 mL | 38-430 mm |
| 342089-0001 | 30 mL | 20-415 mm |
| FKS10304103 | 30 mL | 20-415 mm |
| 342089-0002 | 60 mL | 20-415 mm |
| FIS01-288-34 | 60 mL | 20-415 mm |
| FKS10621294 | 60 mL | 20-415 mm |
| 342089-0004 | 125 mL | 24-415 mm |
| 342089-0008 | 250 mL | 24-415 mm |
| 342089-0016 | 500 mL | 28-415 mm |
| FIS01-288-39 | 500 mL | 28-415 mm |
| FIS01-288-40 | 1000 mL | 38-430 mm |
Minimize biological and particulate contamination with Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Packaging Bottles with Closure. With durable construction and a narrow mouth for easy pouring, these high density polyethylene bottles are produced in a controlled environment, feature excellent chemical resistance and are sterile to 10-6 SAL.
Ordering Information:
Bottles are packed in shrink-wrapped trays with closures assembled.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database