Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Actual product may vary
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Thermo Scientific
Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
342024-0125125 mL38-430 mm
Catalog number 342024-0125
HSN 39233090
Price (SGD)
285.00
Case
Capacity (Metric):
125 mL
Closure Size:
38-430 mm
Price (SGD)
285.00
Case
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:
  • Heavy-walled and durable
  • Shatter-resistant
  • Excellent gas barrier properties
  • Sterile 10-6 SAL
  • Molded in graduations
  • Closure supplied separately
Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
Capacity (English)4 oz.
ColorClear
DescriptionMedia bottles without closure
Diameter (English)2.09 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Filling Lines
MaterialPETG
No. per Case96
No. per Pack24
SterilitySterile
Closure IncludedNo
GraduatedYes
MouthNarrow
ShapeSquare
Fit for PurposeSterile
Height (Metric)110 mm
Product TypeMedia Bottle
Height (English)4.33 in.
Closure Size38-430 mm
Diameter (Metric)53 mm
Unit SizeCase