Check Order Status
Square Cuvettes
Square Cuvettes
Have Questions?
Catalog number
331742-000
Price (SGD)
-
Specifications
Type
Cuvette
Material
Quartz
Path Length
10 mm
Wavelength Range
190 to 1100 nm
For Use With (Equipment)
Helios series spectrophotometers
Shape
Square