Clips and Fasteners for Precision™ Water Baths
Clips and Fasteners for Precision™ Water Baths
Actual product may vary
Thermo Scientific
Clips and Fasteners for Precision™ Water Baths
Use these flask clips in water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Type
31665666 x 250 mL (Model 25), 14 x 250 mL (Model 50) FlasksFlask Clip
316618913 mm to 25 mmClip Fastener
Catalog number 3166566
Price (SGD)
188.00
EA
Add to cart
Capacity (Metric):
6 x 250 mL (Model 25), 14 x 250 mL (Model 50) Flasks
Type:
Flask Clip
Price (SGD)
188.00
EA
Add to cart
Clips and fasteners for use with Thermo Scientific™ Precision™ water baths.
Flask Clips
  • Secure various size flasks to bath platform
  • Stainless steel
  • Each requires one fastener (Cat. No. 3166189 - order separately)
Test Tube Clip
  • Stainless steel
  • Hold 13mm to 25mm tubes
  • Each requires one fastener (Cat. No. 3166189 - order separately)
  • Warranty varies by country, please contact us for details
Specifications
Capacity (Metric)6 x 250 mL (Model 25), 14 x 250 mL (Model 50) Flasks
For Use With (Equipment)For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths.
Shaking MotionReciprocating
TypeFlask Clip
Holds6 x 250 mL (Model 25), 14 x 250 mL (Model 50) Flasks
Unit SizeEA