Check Order Status
Flask Clip For 4 x 500mL (Model 25), 12 x 500mL (Model 50) for Water Baths
Flask Clip For 4 x 500mL (Model 25), 12 x 500mL (Model 50) for Water Baths
Use these flask clips and fasteners in Shallow-Form Shaking Water Baths.
Have Questions?
Catalog number
3166199
Price (SGD)
197.00
EA
Add to cart
Price (SGD)
197.00
EA
Add to cart
Flask Clips
Secure various size flasks to bath platform
Stainless steel
Each requires one fastener (Cat. No. 3166189 - order separately)
Test Tube Clip
Stainless steel
Hold 13mm to 25mm tubes
Each requires one fastener (Cat. No. 3166189 - order separately)
Specifications
For Use With (Equipment)
For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths.
Type
Flask Clip
Holds
4 x 500 mL (Model 25), 12 x 500 mL (Model 50) Flasks