Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL
Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL
Actual product may vary
Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL
Perform low-speed, lipid and phenol extractions, digestions and precipitations with Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer.
Have Questions?
Catalog NumberNominal CapacityMax. Force
3127-0250250 mL4000 x g
Catalog number 3127-0250
Price (SGD)
1,236.00
CS
Add to cart
Nominal Capacity:
250 mL
Max. Force:
4000 x g
Price (SGD)
1,236.00
CS
Add to cart
Perform low-speed, lipid and phenol extractions, digestions and precipitations with Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer. The perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors, they are molded of translucent FEP that resists most acids, bases and organic solvents.
Features:
  • FEP provides excellent chemical resistance
  • Use in temperatures from -100° to +150°C
  • For proper performance, must be filled to 100% of total capacity
  • For added leakproof† asssurance at maximum speeds or when spinning hazardous materials use with PP sealing caps assemblies (DS3131-0038)
Requires:
When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorClear
DescriptionTeflon FEP Bottle with Tefzel ETFE closure
MaterialFEP
No. per Case6
No. per Pack1
Nominal Capacity250 mL
GraduatedNo
Max. Force4000 x g
ShapeRound
Height (Metric)126.5 mm
Product TypeCentrifuge Bottle
Closure MaterialTefzel ETFE
Diameter (Metric)60.0 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS
Frequently asked questions (FAQs)
Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?
Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.