VisionMate™ ST Single-Tube 2D Barcode Reader
VisionMate™ ST Single-Tube 2D Barcode Reader
Actual product may vary
Thermo Scientific
VisionMate™ ST Single-Tube 2D Barcode Reader
Scan single 2D barcodes instantly into your tracking software or other application with this barcode reader, which connects via USB port.
Have Questions?
Catalog NumberConnection Type
3125USB
Catalog number 3125
HSN 84716090
Price (SGD)
3,332.00
EA
Add to cart
Connection Type:
USB
Price (SGD)
3,332.00
EA
Add to cart
Scan 2D barcoded tubes into virtually any application with the compact Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader. Plug the unit into an available USB port and allow the drive to automatically install. Hold any Thermo Scientific™ 2D barcoded storage tube over the scanning window and decode the barcode instantly.
  • Instant read time
  • Simple to install and use
  • For individual sample retrieval
  • Export via comma-separated variable file, Microsoft™ Excel™, XML, COM port, ODBC or TCP socket
Specifications
DescriptionVisionMate ST 2-D Bar Code Reader
Data Transfer MethodComma-separated variable file, Microsoft™ Excel™
Quantity1/Pk., 1/Cs.
TypeBarcode Reader
Connection TypeUSB
Unit SizeEA